Idex 316 Stainless Steel Fitting Assembly, Extra Long, 1/16" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex 316 Stainless Steel Fitting Assembly, Extra Long, 1/16" OD Tubing, 10-32 Coned; 10/PK
Idex | Mfr: 6000-262| Item: 0202231
Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar)!
₹42,100.00INR / Pkg of 10
₹42,100.00INR / Pkg of 10
Buy It Together
Current Item
Idex 316 Stainless Steel Fitting Assembly, Extra Long, 1/16" OD Tubing, 10-32 Coned; 10/PK
Idex 316 Stainless Steel Fitting Assembly, Extra Long, 1/16" OD Tubing, 10-32 Coned; 10/PK
0202231
₹42,100.00INR / Pkg of 10
+ =
Total Price 43600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Ferrule 02022-29.
Key Features
  • Constructed of durable stainless steel
  • Extra Long
  • 10-32 Stainless configuration
  • 10/PK
More About this Item
These 316 stainless steel fitting assemblies include nuts and ferrules are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
Metal fittings should not be used with plastic ports because they can damage the ports.
Specifications
  • Fitting MaterialStainless steel
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.588
  • Max Operating Pressure (PSI)20000
  • Description316 Stainless Steel Fitting Assembly, Extra Long, 1/16" OD Tubing, 10-32 Coned; 10/PK