Hydrion 9400 Plastic pH indicator strips, 5.0 to 9.0, flip top vial packaging
Representative image only.
Hydrion 9400 Plastic pH indicator strips, 5.0 to 9.0, flip top vial packaging
Hydrion | Mfr: 9400| Item: 5920044
Three times the matching area compared to other plastic strips
₹4,500.00INR / Pkg of 100
₹4,500.00INR / Pkg of 100
Buy It Together
Current Item
Hydrion 9400 Plastic pH indicator strips, 5.0 to 9.0, flip top vial packaging
Hydrion 9400 Plastic pH indicator strips, 5.0 to 9.0, flip top vial packaging
5920044
₹4,500.00INR / Pkg of 100
+ + =
Total Price 35200.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Simple, single color match
  • Handy matching color chart
  • Long lasting colors
  • 1 year warranty
More About this Item
Immediate pH testing is available in brilliant, long lasting colors. These direct-reading pH papers give a simple, single color match for all pH values in distinct, bright color separations. Handy flip top vials contain 100 dip sticks and feature a handy matching color chart.
1 Year Warranty
Specifications
  • TestpH
  • Measurement Points5 to 9 pH in 0.5 pH unit increments
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionPlastic pH indicator strips, 5.0 to 9.0, flip top vial packaging