Humboldt H-5815 Accuflame Micro Bunsen Burner with Adjustable Orifice; Lp Gas
Representative image only.
Humboldt H-5815 Accuflame Micro Bunsen Burner with Adjustable Orifice; Lp Gas
Humboldt | Mfr: H-5815| Item: 3613032
₹9,000.00INR / Each
₹9,000.00INR / Each
Buy It Together
Current Item
Humboldt H-5815 Accuflame Micro Bunsen Burner with Adjustable Orifice; Lp Gas
Humboldt H-5815 Accuflame Micro Bunsen Burner with Adjustable Orifice; Lp Gas
3613032
₹9,000.00INR / Each
+ =
Total Price 18200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 3 month warranty
More About this Item
Microburners include adjustable air supply and tapered serrated gas connection. Optional tripod stand 36310-50 supports a 1-L beaker.
3 Month Warranty
Specifications
  • Gas TypeLiquid Propane (LP)
  • BTU Output (per hour)3200
  • Inlet Tube OD (in)0.3125
  • Height (in)3 1/2
  • DescriptionAccuflame Micro Bunsen Burner with Adjustable Orifice; Lp Gas
  • Warranty3 Months