Horiba 3200457719 Ion Standard Solution, Potassium, 2000ppm, 6 x 14 mL

Representative image only.

Horiba 3200457719 Ion Standard Solution, Potassium, 2000ppm, 6 x 14 mL

Horiba | Mfr: 3200457719| Item: 0575465

₹16,400.00INR / Each

₹16,400.00INR / Each

Buy It Together

Current Item

Horiba 3200457719 Ion Standard Solution, Potassium, 2000ppm, 6 x 14 mL
Horiba 3200457719 Ion Standard Solution, Potassium, 2000ppm, 6 x 14 mL
0575465

₹16,400.00INR / Each
+ =
Total Price 16400.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Choose the replacement sensor head and solution kits in low and high concentrations to work with your tester.
Specifications
  • Meter TypeAccessory; Standards and Solutions
  • DescriptionIon Standard Solution, Potassium, 2000ppm, 6 x 14 mL
Documents & Certificates
Reviews
Q&A