Honeywell GK011B11 Salisbury Insulated Glove Kit, Class 0, Black, 11"L, Size 11; One Pair
Representative image only.
Honeywell GK011B11 Salisbury Insulated Glove Kit, Class 0, Black, 11"L, Size 11; One Pair
Honeywell | Mfr: GK011B11| Item: 8643518
Comply with NFPA 70E standard while protecting employees from dangerous electrical injuries
₹66,400.00INR / Each
₹66,400.00INR / Each
Buy It Together
Current Item
Honeywell GK011B11 Salisbury Insulated Glove Kit, Class 0, Black, 11"L, Size 11; One Pair
Honeywell GK011B11 Salisbury Insulated Glove Kit, Class 0, Black, 11"L, Size 11; One Pair
8643518
₹66,400.00INR / Each
+ =
Total Price 82000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
insulating rubber glove, leather protector glove, and storage bag.
More About this Item
Get everything you need to protect yourself and keep your insulating rubber gloves in top working condition. Kits include a pair of insulating rubber gloves; the appropriate leather protector that protects the glove from cuts, abrasions, and punctures; and a storage bag which keeps the glove flat, preventing folds and creases that strain the rubber, shortening the life of the glove.
Note: In accordance with NFPA 70E, anything over 50 volts requires insulating rubber gloves.
Specifications
  • MaterialRubber
  • Size11
  • Length (in)11
  • Length (cm)27.9
  • ColorBlack
  • ComplianceClass 0
  • SterileNo
  • DescriptionInsulated Glove Kit, Class 0, Black, 11"L, Size 11; One Pair