Salisbury by Honeywell Leather Protector Gloves
Leather protector gloves should always be worn over Insulating Rubber Gloves to provide mechanical protection against cuts, abrasions, and punctures. Made from top grain goat leather, the protectors increase the life of your rubber gloves.
15,600.00INR / Pair of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
Honeywell ILPG109 Salisbury Goat Skin Leather Protector Glove, 10" length, size 9 8643273 Mfr # ILPG10/9
COMPARE
10 9
₹15,600.00INR / Pair of 1
Honeywell ILPG1010 Salisbury Goat Skin Leather Protector Glove, 10" length, size 10 8643275 Mfr # ILPG10/10
COMPARE
10 10
₹15,600.00INR / Pair of 1
Honeywell ILPG1011 Salisbury Goat Skin Leather Protector Glove, 10" length, size 11 8643277 Mfr # ILPG10/11
COMPARE
10 11
₹15,600.00INR / Pair of 1
Honeywell ILPG1012 Salisbury Goat Skin Leather Protector Glove, 10" length, size 12 8643279 Mfr # ILPG10/12
COMPARE
10 12
₹15,600.00INR / Pair of 1