High-Tech Conversions FS-NT1-712.50 Cleanroom wipes, pre-saturated nonwoven poly-cellulose, 50% isopropyl alcohol/50% deionized water, 7" x 12", 900/CS
Representative image only.
High-Tech Conversions Mfr # FS-NT1-712.50 – Item # WW-15981-12
Never worry again about fiber contamination from these cleanroom wipes
  • Virtually free of particle generation
  • Choose from ISO Class 3 or Class 5 compliance
  • No residual lint for controlled environment use
  • 2 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNonwoven poly-cellulose
  • ComplianceISO Class 5
  • Width (in)12
  • Length (in)7
  • Width (cm)30.5
  • Length (cm)17.8
  • DescriptionCleanroom wipes, pre-saturated nonwoven poly-cellulose, 50% isopropyl alcohol/50% deionized water, 7\" x 12\", 900/CS
  • Warranty2 year
Two year warranty
Key Features
  • Virtually free of particle generation
  • Choose from ISO Class 3 or Class 5 compliance
  • No residual lint for controlled environment use
  • 2 year warranty
More About this Item
These cleanroom wipes feature four clean-cut edges to eliminate fiber contamination, this also allows for no long strands or loose fibers typically found on perforated roll-type wipes. These presaturated wipes are available in different concentrations of an isopropyl alcohol (IPA) and deionized (DI) water mix, a range of sizes, as well as materials. Models 15981-09 and 15981-10 are made of polypropylene for a high level of absorbency. Models 15981-11, 15981-12, and 15981-13 are all made of nonwoven polycellulose, allowing them to be tough and abrasion resistant for applications in which a durable wipe is required. Model 15981-14 features a tough, interlocking kitted polyester construction, making it a heavy-weight, high-absorbent wipe ideal for heavy duty applications.