HF Scientific 52050 Hach ProCal Primary Standard for Process Turbidimeter 1720, 0.0 NTU
Representative image only.
HF Scientific 52050 Hach ProCal Primary Standard for Process Turbidimeter 1720, 0.0 NTU
HF Scientific | Mfr: 52050| Item: 9990034
Instrument specific kits and standards ensure maximum accuracy
₹23,700.00INR / Each
₹23,700.00INR / Each
Buy It Together
Current Item
HF Scientific 52050 Hach ProCal Primary Standard for Process Turbidimeter 1720, 0.0 NTU
HF Scientific 52050 Hach ProCal Primary Standard for Process Turbidimeter 1720, 0.0 NTU
9990034
₹23,700.00INR / Each
+ + =
Total Price 265300.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
  • 1 year warranty
More About this Item
ProCal Primary Turbidity Standards are formulated to provide instrument-specific accuracy with a safe, non toxic formulation, and offer improvements in stability over polymer and formazin alternatives, two-year shelf life. U.S. EPA-approved to meet NPDES and NPDWR compliance reporting requirements.
One 1-L bottle
1 Year Warranty
Specifications
  • NTU Value0.0
  • Volume (mL)1000
  • Volume (oz)33.8
  • Use withHach 1720 Process Turbidimeters
  • DescriptionHach ProCal Primary Standard for Process Turbidimeter 1720, 0.0 NTU