HF Scientific 39957 Calibration Kit for MicroTOL and MTOL+ Turbidimeters, 0.02, 10 and 1000 NTU
Representative image only.
HF Scientific 39957 Calibration Kit for MicroTOL and MTOL+ Turbidimeters, 0.02, 10 and 1000 NTU
HF Scientific | Mfr: 39957| Item: 9954350
Optimize water analysis
₹194,000.00INR / Each
₹194,000.00INR / Each
Buy It Together
Current Item
HF Scientific 39957 Calibration kit for MicroTOL and MTOL+ Turbidimeters, 0.02, 10 and 1000 NTU
HF Scientific 39957 Calibration kit for MicroTOL and MTOL+ Turbidimeters, 0.02, 10 and 1000 NTU
9954350
₹194,000.00INR / Each
+ + + + =
Total Price 807300.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Keep your meter running smoothly and accurately with these parts and accessories.
1 Year Warranty
Specifications
  • Compatible WithHF Scientific Micro 100 turbidimeter, 99543-00 through99543-15
  • DescriptionCalibration set for turbidity meters item numbers 99543-00 and -05