HF Scientific 29845 PrimeTime Calibration Kit, (0.02, 10, 1000 NTU)
Representative image only.
HF Scientific 29845 PrimeTime Calibration Kit, (0.02, 10, 1000 NTU)
HF Scientific | Mfr: 39845| Item: 9954360
Optimize water analysis
₹105,000.00INR / Each
₹105,000.00INR / Each
Buy It Together
Current Item
HF Scientific 29845 PrimeTime Calibration Kit, (0.02, 10, 1000 NTU)
HF Scientific 29845 PrimeTime Calibration Kit, (0.02, 10, 1000 NTU)
9954360
₹105,000.00INR / Each
+ + + + =
Total Price 718300.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Choose from a variety of accessories
  • Replacement calibration set for MicroTPW Handheld Turbidimeter 99543-20
  • Includes 0.02, 10, and 1000 NTU standards
  • 1 year warranty
More About this Item
Keep your meter running smoothly and accurately with these parts and accessories.
1 Year Warranty
Specifications
  • Compatible WithHF Scientific MicroTPW turbidimeter, 99543-00 through99543-15
  • DescriptionPrimeTime Calibration Kit, (0.02, 10, 1000 NTU)