Hamilton Removable Syringe Needle, Large Hub, Blunt Tip, 22 G; 6/Pk
Representative image only.
Hamilton Removable Syringe Needle, Large Hub, Blunt Tip, 22 G; 6/Pk
Hamilton | Mfr: 7780-04| Item: 2570264
Ideal for chromatography
₹13,200.00INR / Pkg of 6
₹13,200.00INR / Pkg of 6
Buy It Together
Current Item
Hamilton Removable Syringe Needle, Large Hub, Blunt Tip, 22 G; 6/Pk
Hamilton Removable Syringe Needle, Large Hub, Blunt Tip, 22 G; 6/Pk
2570264
₹13,200.00INR / Pkg of 6
+ + =
Total Price 85900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of durable 304 stainless steel
  • Autoclavable
  • Needle point style 3 features a blunt needle point for use with HPLC injection valves and for sample pipetting
  • Large hub removable (RN) needles can be used with removable needle syringes 250 μL and larger, or SampleLock syringes
  • 1 year warranty
More About this Item
Hamilton® removable syringe needles offer the benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • Syringe TypeLarge hub removable needle
  • Needle Tip StyleBlunt
  • Needle Tip Number3
  • Needle Gauge22
  • Needle Length (in)2
  • Material304 stainless steel
  • DescriptionRemovable Syringe Needle, Large Hub, Blunt Tip, 22 G; 6/Pk