Hamilton Removable Syringe Needle, Large Hub, Beveled Tip, 22 G; 6/Pk
Representative image only.
Hamilton Removable Syringe Needle, Large Hub, Beveled Tip, 22 G; 6/Pk
Hamilton | Mfr: 7779-01| Item: 0793886
Ideal for chromatography
₹14,200.00INR / Pkg of 6
₹14,200.00INR / Pkg of 6
Buy It Together
Current Item
Hamilton Removable Syringe Needle, Large Hub, Beveled Tip, 22 G; 6/Pk
Hamilton Removable Syringe Needle, Large Hub, Beveled Tip, 22 G; 6/Pk
0793886
₹14,200.00INR / Pkg of 6
+ + =
Total Price 82000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of durable 304 stainless steel
  • Autoclavable
  • Needle point style 2 features a sharp, beveled, curved, non-coring needle point recommended for piercing septa
  • Large hub removable (RN) needles can be used with removable needle syringes 250 μL and larger, or SampleLock syringes
  • 1 year warranty
More About this Item
Hamilton® removable syringe needles offer the benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • Syringe TypeLarge hub removable needle
  • Needle Tip Number2
  • Needle Tip StyleSharp, beveled, curved, non-coring
  • Needle Gauge22
  • Needle Length (in)2
  • Material304 stainless steel
  • DescriptionRemovable Syringe Needle, Large Hub, Beveled Tip, 22 G; 6/Pk