Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 250 µL
Representative image only.
Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 250 µL
Hamilton | Mfr: 80701| Item: 0793824
Achieve precise liquid delivery
₹22,100.00INR / Each
₹22,100.00INR / Each
Buy It Together
Current Item
Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 250 µL
Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 250 µL
0793824
₹22,100.00INR / Each
+ + =
Total Price 94800.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of borosilicate glass
  • Syringe features a 1000 psi max pressure
  • 18 month warranty
More About this Item
Hamilton® 700-series microliter syringes are designed to solve the general liquid handling requirements of manufacturing and research laboratories. Syringes are used for manual and automated GC and HPLC injections and for everyday applications that require accurate liquid measuring and dispending.
Choose from cemented needle, removable needle, or Luer tip syringes. Syringes with luer tip connections do not include a needle, order needles separately.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
18 Month Warranty
Specifications
  • Syringe TypeLuer Tip
  • Syringe Capacity (µL)250
  • Needle IncludedNo
  • Needle GaugeNeedle not included
  • MaterialSST, borosilicate glass
  • Outside Diameter (OD) (mm)7.75
  • DescriptionMicroliter Low-Volume Syringe, Luer Tip, No Needle, 250 µL
  • Warranty18 months