Hamilton Gastight® Instrument Syringe, Borosilicate Glass, 500 µL, PTFE Luer Lock
Representative image only.
Hamilton Gastight® Instrument Syringe, Borosilicate Glass, 500 µL, PTFE Luer Lock
Hamilton | Mfr: 81222| Item: 0793790
Gastight® syringe designed to fit different instruments
₹17,000.00INR / Each
₹17,000.00INR / Each
Buy It Together
Current Item
Hamilton Gastight® Instrument Syringe, Borosilicate Glass, 500 µL, PTFE Luer Lock
Hamilton Gastight® Instrument Syringe, Borosilicate Glass, 500 µL, PTFE Luer Lock
0793790
₹17,000.00INR / Each
+
Cole-Parmer PTFE Stir Bar, Plain, 13 x 8mm
0477020
₹2,100.00INR / Each
=
Total Price 19100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of borosilicate glass
  • 1 year warranty
More About this Item
Designed to fit into, or with, various different instruments. Fits drive arm mechanisms, like those found on the Hamilton 500 and 1000 Series diluters/dispensers.
1 Year Warranty
Specifications
  • Syringe TypePTFE Luer Lock
  • Syringe Capacity (µL)500
  • Needle IncludedNo
  • MaterialBorosilicate glass, PTFE
  • SterileNo
  • Outside Diameter (OD) (mm)7.75
  • DescriptionInstrument Syringe, Borosilicate Glass, 500 µL, PTFE Luer Lock