Hach TNT839 TNTplus® Spectrophotometer Chemistry, Low Range Nitrite, 0.015 to 0.6 mg/L; 25/PK
Representative image only.
Hach TNT839 TNTplus® Spectrophotometer Chemistry, Low Range Nitrite, 0.015 to 0.6 mg/L; 25/PK
Hach | Mfr: TNT839| Item: 9951532
Perform common methods with these pre-made reagent tests
₹27,700.00INR / Pkg of 25
₹27,700.00INR / Pkg of 25
Buy It Together
Current Item
Hach TNT839 TNTplus® Spectrophotometer Chemistry, Low Range Nitrite, 0.015 to 0.6 mg/L; 25/PK
Hach TNT839 TNTplus® Spectrophotometer Chemistry, Low Range Nitrite, 0.015 to 0.6 mg/L; 25/PK
9951532
₹27,700.00INR / Pkg of 25
+ +
Hach DR3900 Benchtop Visible Spectrophotometer
9951500
₹3,103,000.00INR / Each
+
Hach 2667200 Iron, Mid Range Color Disc Test Kit
0554565
₹47,800.00INR / Each
=
Total Price 3203300.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Measures: Low Range Nitrite, 0.015 to 0.6 mg/L
More About this Item
Dozens of options to cover most applications!
Specifications
  • MaterialLow Range Nitrite, 0.015 to 0.6 mg/L
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, Low Range Nitrite, 0.015 to 0.6 mg/L; 25/PK