Hach TNT836 TNTplus® Spectrophotometer Chemistry, High Range Nitrate, 5 to 35 mg/L Nitrogen; 25/PK
Representative image only.
Hach TNT836 TNTplus® Spectrophotometer Chemistry, High Range Nitrate, 5 to 35 mg/L Nitrogen; 25/PK
Hach | Mfr: TNT836| Item: 9951531
Perform common methods with these pre-made reagent tests
Call For Price
Call For Price
Buy It Together
Current Item
Hach TNT836 TNTplus® Spectrophotometer Chemistry, High Range Nitrate, 5 to 35 mg/L Nitrogen; 25/PK
Hach TNT836 TNTplus® Spectrophotometer Chemistry, High Range Nitrate, 5 to 35 mg/L Nitrogen; 25/PK
9951531
Call For Price
+
Hach 2667200 Iron, Mid Range Color Disc Test Kit
0554565
₹47,800.00INR / Each
+
Hach DR3900 Benchtop Visible Spectrophotometer
9951500
₹3,103,000.00INR / Each
+ =
Total Price 3175600.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Measures: High Range Nitrate, 5 to 35 mg/L Nitrogen
More About this Item
Dozens of options to cover most applications!
Specifications
  • MaterialHigh Range Nitrate, 5 to 35 mg/L Nitrogen
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, High Range Nitrate, 5 to 35 mg/L Nitrogen; 25/PK