Hach TNT833 TNTplus® Spectrophotometer Chemistry, Ultra High Range Ammonia, 47 to 130 mg/L NH3-N; 25/PK
Representative image only.
Hach TNT833 TNTplus® Spectrophotometer Chemistry, Ultra High Range Ammonia, 47 to 130 mg/L NH3-N; 25/PK
Hach | Mfr: TNT833| Item: 9951529
Perform common methods with these pre-made reagent tests
Call For Price
Call For Price
Buy It Together
Current Item
Hach TNT833 TNTplus® Spectrophotometer Chemistry, Ultra High Range Ammonia, 47 to 130 mg/L NH3-N; 25/PK
Hach TNT833 TNTplus® Spectrophotometer Chemistry, Ultra High Range Ammonia, 47 to 130 mg/L NH3-N; 25/PK
9951529
Call For Price
+ +
Hach DR3900 Benchtop Visible Spectrophotometer
9951500
₹3,103,000.00INR / Each
+
Hach 2667200 Iron, Mid Range Color Disc Test Kit
0554565
₹47,800.00INR / Each
=
Total Price 3175600.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Measures: Ultra High Range Ammonia, 47 to 130 mg/L NH3-N
More About this Item
Dozens of options to cover most applications!
Specifications
  • MaterialUltra High Range Ammonia, 47 to 130 mg/L NH3-N
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, Ultra High Range Ammonia, 47 to 130 mg/L NH3-N; 25/PK