Hach TNT811 TNTplus® Spectrophotometer Chemistry, High Range TOC, 30 to 300 mg/L; 25/PK
Representative image only.
Hach TNT811 TNTplus® Spectrophotometer Chemistry, High Range TOC, 30 to 300 mg/L; 25/PK
Hach | Mfr: TNT811| Item: 9951512
Perform common methods with these pre-made reagent tests
₹251,000.00INR / Pkg of 25
₹251,000.00INR / Pkg of 25
Buy It Together
Current Item
Hach TNT811 TNTplus® Spectrophotometer Chemistry, High Range TOC, 30 to 300 mg/L; 25/PK
Hach TNT811 TNTplus® Spectrophotometer Chemistry, High Range TOC, 30 to 300 mg/L; 25/PK
9951512
₹251,000.00INR / Pkg of 25
+ +
Hach DR3900 Benchtop Visible Spectrophotometer
9951500
₹3,103,000.00INR / Each
+
Hach 2667200 Iron, Mid Range Color Disc Test Kit
0554565
₹47,800.00INR / Each
=
Total Price 3426600.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Measures: High Range TOC, 30 to 300 mg/L
More About this Item
Dozens of options to cover most applications!
Specifications
  • MaterialHigh Range TOC, 30 to 300 mg/L
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, High Range TOC, 30 to 300 mg/L; 25/PK