Hach TNT810 TNTplus® Spectrophotometer Chemistry, Low Range TOC, 1.5 to 30 mg/L; 25/PK
Representative image only.
Hach TNT810 TNTplus® Spectrophotometer Chemistry, Low Range TOC, 1.5 to 30 mg/L; 25/PK
Hach | Mfr: TNT810| Item: 9951511
Perform common methods with these pre-made reagent tests
₹251,000.00INR / Pkg of 25
₹251,000.00INR / Pkg of 25
Buy It Together
Current Item
Hach TNT810 TNTplus® Spectrophotometer Chemistry, Low Range TOC, 1.5 to 30 mg/L; 25/PK
Hach TNT810 TNTplus® Spectrophotometer Chemistry, Low Range TOC, 1.5 to 30 mg/L; 25/PK
9951511
₹251,000.00INR / Pkg of 25
+
Hach 2667200 Iron, Mid Range Color Disc Test Kit
0554565
₹47,800.00INR / Each
+
Hach DR3900 Benchtop Visible Spectrophotometer
9951500
₹3,103,000.00INR / Each
+ =
Total Price 3426600.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Measures: Low Range TOC, 1.5 to 30 mg/L
More About this Item
Dozens of options to cover most applications!
Specifications
  • MaterialLow Range TOC, 1.5 to 30 mg/L
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, Low Range TOC, 1.5 to 30 mg/L; 25/PK