Hach HQ2100 Portable Multiparameter with Low-Maintenance Gel pH Electrode, 5 m Rugged Cable

Representative image only.

Hach HQ2100 Portable Multiparameter with Low-Maintenance Gel pH Electrode, 5 m Rugged Cable

Hach | Mfr: LEV015.53.21003| Item: 5900518

A robust and intuitive multiparameter meter

₹827,000.00INR / Each

₹827,000.00INR / Each

Buy It Together

Current Item

Hach HQ2100 Portable Multiparameter with Low-Maintenance Gel pH Electrode, 5 m Rugged Cable
Hach HQ2100 Portable Multiparameter with Low-Maintenance Gel pH Electrode, 5 m Rugged Cable
5900518

₹827,000.00INR / Each
+
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter
5900500

₹274,000.00INR / Each
+
Hach HQ1140  Portable Dedicated Conductivity/TDS Meter
5900508

₹293,000.00INR / Each
+
Hach HQ1130  Portable Dedicated Dissolved Oxygen Meter
5900505

₹293,000.00INR / Each
+ +
Hach LDO10101 IntelliCAL LDO Lab Probe, 1 m cable
5900457

₹350,000.00INR / Each
=
Total Price 2128000.00 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Meter, PHC101 rugged field pH electrode (59004-74), rechargeable lithium-ion battery, USB power adapter, and USB cable.

Key Features

  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 10,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
  • 5 m rugged cable low maintenance gel pH electrode
  • 1 year warranty

More About this Item

Hach portable HQ meters offer a versatile range of measurements, including pH, conductivity, total dissolved solids (TDS), salinity, dissolved oxygen (DO), oxidation reaction potential (ORP), and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.

The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 536 x 335 Mono-TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.

1 Year WarrantyConformite Europeenne
Specifications
  • Meter TypepH, conductivity, TDS, salinity, DO, ORP
  • Display type536x336 Mono-TFT
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.02 pH
  • Buffer RecognitionIUPAC, USA, DIN, Custom
  • CalibrationUp to 3 points
  • Min mV (mV)-1200
  • mV Resolution0.1 mV
  • mV AccuracyProbe specific
  • Min Conductivity0.01 µS/cm
  • Max Conductivity200 mS/cm
  • Conductivity Resolution0.01 µS/cm to 0.1 mS/cm upon selected measuring range
  • Conductivity AccuracyProbe specific
  • Min TDS0.01 g/L
  • Max TDS50 g/L
  • TDS Resolution0.01 mg/L up - 0.1 g/L (range dependent)
  • TDS AccuracyProbe specific
  • Min Salinity42 ppt or %
  • Max Salinity0 ppt or %
  • Salinity Resolution0.01 (range dependent)
  • Salinity AccuracyProbe specific
  • Min Dissolved Oxygen0.0 mg/L
  • Max Dissolved Oxygen27 mg/L
  • Dissolved Oxygen Resolution0.1 mg/L
  • Dissolved Oxygen AccuracyProbe specific
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature Resolution 0.1°C
  • Temperature Accuracy±0.3 °C (±0.54 °F)
  • Temperature compensationATC
  • OutputUSB
  • Data Logging (points)Yes
  • Memory Size10,000 data points
  • WaterproofIP67
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Battery1 rechargeable Lithium-ion (included)
  • Length (cm)22
  • Width (cm)9.7
  • Height (cm)6.3
  • Width (in)3 13/16
  • Length (in)8 21/32
  • Height (in)2 31/64
  • Number of Positions1
  • NIST Calibration IncludedNo
  • Probe Included5 m cable low maintenance gel pH electrode
  • DescriptionPortable Multi-Meter with Low Maintenance Gel pH Electrode, 5 m Rugged Cable
  • Warranty1 year
Reviews
Q&A