Glas-Col 099C L15 Tissue Homogenizing System Flex-Grip Chuck, 6 mm dia pestle
Representative image only.
Glas-Col 099C L15 Tissue Homogenizing System Flex-Grip Chuck, 6 mm dia pestle
Glas-Col | Mfr: 099C L15| Item: 0436951
Complete your setup with chucks, flexible drive shaft, and lab stand
₹44,600.00INR / Each
₹44,600.00INR / Each
Buy It Together
Current Item
Glas-Col 099C L15 Tissue Homogenizing System Flex-Grip Chuck, 6 mm dia pestle
Glas-Col 099C L15 Tissue Homogenizing System Flex-Grip Chuck, 6 mm dia pestle
0436951
₹44,600.00INR / Each
+ =
Total Price 367600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 3 month warranty
More About this Item
Flex-Grip Chucks are flexible, self-gripping slip clutches. Connections in the chuck accept glass pestles directly — no need for tubing connector. Use with pestles with glass shafts or glass stirrer rods measuring 6 mm.
3 Month Warranty
Specifications
  • DescriptionTissue Homogenizing System Flex-Grip Chuck, 6 mm dia pestle