GF Signet 3-9900.394 Universal Transmitter Direct Conductivity Module
Representative image only.
GF Signet 3-9900.394 Universal Transmitter Direct Conductivity Module
GF Signet | Mfr: 3-9900.394| Item: 5656016
Optimize performance of your GF+ Signet Transmitter/Controller
₹62,600.00INR / Each
₹62,600.00INR / Each
Buy It Together
Current Item
GF Signet 3-9900.394 Universal Transmitter Direct Conductivity Module
GF Signet 3-9900.394 Universal Transmitter Direct Conductivity Module
5656016
₹62,600.00INR / Each
+ + + =
Total Price 336200.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a wide range of replacement parts and accessories
  • Required for Conductivity Measurements on the 9900 Transmitter
More About this Item
Use modules, sensors, and RC filters to expand the controller's capabilities. Adapt your controller to your existing system with a variety of mounting options.
A complete conductivity/resistivity system requires a 9900 Transmitter (56560-12 or 56560-14), this Direct Conductivity/Resistivity Module, and a Conductivity cell .
Conformite Europeenne
Specifications
  • Control TypeOn/Off
  • DescriptionUniversal Transmitter Direct Conductivity Module