General Tools DT4208SD 12-Channel Temperature Recorder with Sd Card Data Logging
Representative image only.
General Tools DT4208SD 12-Channel Temperature Recorder with Sd Card Data Logging
General Tools | Mfr: DT4208SD| Item: 3780308
Accurately measures 12 channels of temperature independently
₹300,000.00INR / Each
for an additional
₹36,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹300,000.00INR / Each
Buy It Together
Current Item
General Tools DT4208SD 12-Channel Temperature Recorder with Sd Card Data Logging
General Tools DT4208SD 12-Channel Temperature Recorder with Sd Card Data Logging
3780308
₹300,000.00INR / Each
+ + =
Total Price 343900.00 ADD 3 ITEMS TO CART
Product Description
Includes
two Type K beaded wire temperature probes, 2G SD card, and hard carrying case.
Key Features
  • Records data onto an SD card in Excel® format for easy transfer to a PC for analysis
  • Datalogging capability up to 20,000 records using a 2G SD card
  • Manual store and recall of up to 99 records
  • 1 year warranty
More About this Item
The 12-channel meter has automatic temperature compensation and is designed to work with the six most popular thermocouple types: J, K, T, E, S, and R. Data logging capability up to 20,000 records using a 2G SD card, with user-programmable sampling rate from one second up to one hour.
Data is recorded onto a standard SD memory card instead of a built-in memory, offering unlimited data storage. The removable SD card is easy to transport and insert into a computer’s memory card slot or SD card reader for upload. Data is provided in Excel, allowing you to easily analyze the information without the need for additional software. Data can also be streamed directly to a computer using the RS-232 port on the meter.
Additional meter features include a large 2-1/2” backlit display, min/max, hold, offset adjustment, loop data recording, sampling interval, date-and-time stamp, selectable units of measure, period or comma decimal point division, auto power off, and low-battery indication. The unit operates on either battery or AC power.
1 Year Warranty
Specifications
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels2
  • Types Of Probes AcceptedType E, Type J, Type K, Type R, Type S, Type T
  • Min Temperature (° F)-148
  • Max Temperature (° F)2372
  • Min Temperature (° C)-100
  • Max Temperature (° C)1300
  • Accuracy±0.4% of reading
  • Resolution0.1°
  • OutputSD card
  • Data LoggingYes
  • Memory Capacity20000 data points
  • Display TypeLCD
  • No. of Probe(s) Included12
  • Battery8 AA (not included)
  • Description12-Channel Temperature Recorder with Sd Card Data Logging