Gardco EZ5/C series viscosity cup #5 with certificate

Representative image only.

Gardco EZ5/C series viscosity cup #5 with certificate

Gardco | Mfr: EZ5/C| Item: 0870042

An easy and economical way to measure viscosity

₹129,000.00INR / Each

for an additional
₹28,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹129,000.00INR / Each

Buy It Together

Current Item

Gardco EZ5/C series viscosity cup #5 with certificate
Gardco EZ5/C series viscosity cup #5 with certificate
0870042

₹129,000.00INR / Each
+
InnoCal NIST-Traceable Calibration; Viscosity Cup
1710702

₹28,000.00INR / Each
+ + =
Total Price 379300.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • ±3% production tolerance

More About this Item

Easily measure viscosity using an EZ viscosity cup and a stopwatch. Simply fill the cup by immersing it in the test liquid. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. Each cup is supplied with a conversion table to determine viscosity in centistokes (cSt) from the elapsed time.
Each stainless steel cup meets dimensions as specified in ASTM D 4212 standards.

NIST-traceable models conform to MIL STD 5662A, ANSI/NCSL 2540-1, and are traceable to NIST-standard oils. NIST-traceable models include a certificate verifying traceability to NIST standards.

Specifications
  • Viscosity161 to 1401 centistokes
  • Sample Volume Needed (mL)46.5
  • Cup Number5
  • Nist-TraceableYes
  • Descriptionseries viscosity cup #5 with certificate
Reviews
Q&A