Gardco EZ3 Series Viscosity Cup #3, 64 to 596 Centistokes

Representative image only.

Gardco EZ3 Series Viscosity Cup #3, 64 to 596 Centistokes

Gardco | Mfr: EZ3| Item: 0870020

An easy and economical way to measure viscosity

₹55,300.00INR / Each

for an additional
₹28,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹55,300.00INR / Each

Buy It Together

Current Item

Gardco EZ3 Series Viscosity Cup #3, 64 to 596 Centistokes
Gardco EZ3 Series Viscosity Cup #3, 64 to 596 Centistokes
0870020

₹55,300.00INR / Each
+
Gardco VI-3403 Standard Ford Dip Viscosity Cup, orifice #3
0870900

₹142,000.00INR / Each
+ +
InnoCal NIST-Traceable Calibration; Viscosity Cup
1710702

₹28,000.00INR / Each
=
Total Price 299700.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • ±3% production tolerance

More About this Item

Easily measure viscosity using an EZ viscosity cup and a stopwatch. Simply fill the cup by immersing it in the test liquid. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. Each cup is supplied with a conversion table to determine viscosity in centistokes (cSt) from the elapsed time.
Each stainless steel cup meets dimensions as specified in ASTM D 4212 standards.

Specifications
  • Viscosity64 to 596 centistokes
  • Sample Volume Needed (mL)46.5
  • Cup Number3
  • Nist-TraceableNo
  • DescriptionSeries Viscosity Cup #3, 64 to 596 Centistokes
Reviews
Q&A