Extech 480403 Phase Sequence and Motor Rotation Tester
Representative image only.
Extech 480403 Phase Sequence and Motor Rotation Tester
Extech | Mfr: 480403| Item: 2682350
Check both phase sequence and motor rotation with one meter
₹51,000.00INR / Each
for an additional
₹36,600.00INR / Each
for an additional
₹48,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹51,000.00INR / Each
Buy It Together
Current Item
Extech 480403 Phase Sequence and Motor Rotation Tester
Extech 480403 Phase Sequence and Motor Rotation Tester
2682350
₹51,000.00INR / Each
+
InnoCal NIST-Traceable Calibration, Hipot Tester
1710128
₹36,600.00INR / Each
+
Extech 382252 Earth ground Resistance Tester Kit
2004208
₹85,300.00INR / Each
=
Total Price 172900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Indicates rotation direction of the motor
  • Determine rotation of a motor without contact
  • Ensures that motor does not get damaged from incorrect wiring
  • 120 to 400VAC rated test range for testing phase orientation of three phase power sources over 2 to 400Hz frequency range
  • LEDs indicate phase orientation (clockwise or counter-clockwise) and whether each of the three phases is live
  • Durable housing is double molded
  • Easy to open alligator clips with wide jaws
  • Complete with cable and three large color coded alligator clips, pouch case, and 9V battery
  • 1 year warranty
1 Year Warranty
Specifications
  • DC Min (Volts)120
  • DC Max (Volts)400
  • AC Min (Volts)120
  • AC Max (Volts)400
  • Safety RatingCAT III 600V, EN 61010-1
  • DescriptionPhase Sequence and Motor Rotation Tester