Environmental Express TKN6011 TKN Digestion Tube, Non-Threaded; 1/Ea
Representative image only.
Environmental Express TKN6011 TKN Digestion Tube, Non-Threaded; 1/Ea
Environmental Express | Item: 3520573
Optimize your TKN analysis for better results
₹7,700.00INR / Each
₹7,700.00INR / Each
Buy It Together
Current Item
Environmental Express TKN6011 TKN Digestion Tube, Non-Threaded; 1/Ea
Environmental Express TKN6011 TKN Digestion Tube, Non-Threaded; 1/Ea
3520573
₹7,700.00INR / Each
+ + =
Total Price 58800.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Interchangeable glassware works with SimpleDist and TKN HotBLock systems
  • Maximum heat transfer with flat-bottom, borosilicate glassware
  • Ideal for reflux and digestion methods
  • Model number TKN6011
More About this Item
Teardrop stoppers are ideal for the capture and condesation of digestion fumes, preventing cross contamination (sold separately).
Specifications
  • Capacity (mL)50
  • DescriptionNon-Threaded Digestion Tube, EA