Environmental Express O-Ring, Calibration Adapter, Aluminum Cyclone; 5/Pk
Representative image only.
Environmental Express O-Ring, Calibration Adapter, Aluminum Cyclone; 5/Pk
Environmental Express | Item: 6006191
Help ensure flow rate accuracy
₹1,400.00INR / Pkg of 5
₹1,400.00INR / Pkg of 5
Buy It Together
Current Item
Environmental Express O-Ring, Calibration Adapter, Aluminum Cyclone; 5/Pk
Environmental Express O-Ring, Calibration Adapter, Aluminum Cyclone; 5/Pk
6006191
₹1,400.00INR / Pkg of 5
+ =
Total Price 9400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Fits on both 25- and 37-mm aluminum cyclones
  • Ensure a tight fit with an O-ring
More About this Item
Use a calibration adapter to ensure the flow rate accuracy. Place the adapter over the stem of a 25- or 37-mm cyclone then connect to a flow calibrator using a 1/4" ID tubing. This allows an actual measurement as close to the cyclone inlet as possible for the most accurate reading.
Specifications
  • Compatible WithAluminum cyclone calibration adapter
  • DescriptionZefon O-Ring, Calibration Adapter, Aluminum Cyclone; 5/PK