Environmental Express Calibration Adapter, Aluminum Cyclone
Representative image only.
Environmental Express Calibration Adapter, Aluminum Cyclone
Environmental Express | Item: 6006190
Help ensure flow rate accuracy
₹8,000.00INR / Each
₹8,000.00INR / Each
Buy It Together
Current Item
Environmental Express Calibration Adapter, Aluminum Cyclone
Environmental Express Calibration Adapter, Aluminum Cyclone
6006190
₹8,000.00INR / Each
+ =
Total Price 9400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Fits on both 25- and 37-mm aluminum cyclones
  • Help ensure flow rate accuracy
More About this Item
Use a calibration adapter to ensure the flow rate accuracy. Place the adapter over the stem of a 25- or 37-mm cyclone then connect to a flow calibrator using a 1/4" ID tubing. This allows an actual measurement as close to the cyclone inlet as possible for the most accurate reading.
Specifications
  • MaterialAluminum
  • Compatible With25- and 37-mm aluminum cyclones
  • DescriptionZefon Calibration Adapter, Aluminum Cyclone