Environmental Express HotBlock Pro Temperature Probe, External
Representative image only.
Environmental Express HotBlock Pro Temperature Probe, External
Environmental Express | Item: 5320162
Obtain real-time temperature readings from your digestion cups
₹146,000.00INR / Each
for an additional
₹30,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹146,000.00INR / Each
Buy It Together
Current Item
Environmental Express HotBlock Pro Temperature Probe, External
Environmental Express HotBlock Pro Temperature Probe, External
5320162
₹146,000.00INR / Each
+ =
Total Price 188700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Temperature probe allows you to keep your reaction temperature steady
  • The holder makes temperature readings easy
  • 1 year warranty
More About this Item
The HotBlock Pro controller will simultaneously display the actual cup temperature, as well as the set temperature, to give you a better feel for your reactions.
One controller and up to two blocks are required for a full system; temperature probe is optional, but suggested.
1 Year Warranty
Specifications
  • Probe TypeThermocouple
  • DescriptionHotBlock Pro Temperature Probe, External