Environmental Express Flipmate™ 50 Filtration Systems, Polypropylene, 0.7um, Glass Fiber Filter with Pre-Filter; 200/pk
Representative image only.
Environmental Express Flipmate™ 50 Filtration Systems, Polypropylene, 0.7um, Glass Fiber Filter with Pre-Filter; 200/pk
Environmental Express | Item: 5310009
Convenient, economical method for filtration
₹72,700.00INR / Pkg of 200
₹72,700.00INR / Pkg of 200
Buy It Together
Current Item
Environmental Express Flipmate™ 50 Filtration Systems, Polypropylene, 0.7um, Glass Fiber Filter with Pre-Filter; 200/pk
Environmental Express Flipmate™ 50 Filtration Systems, Polypropylene, 0.7um, Glass Fiber Filter with Pre-Filter; 200/pk
5310009
₹72,700.00INR / Pkg of 200
+
Environmental Express Vacuum Manifold, 12-Pl
5320519
Call For Price
+ =
Total Price 98100.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Produce quality results with clean samples
  • Results within seconds
More About this Item
The FlipMate system easily filters the sample with the use of two threaded digestion cups, a filter assembly, vacuum tubing, and vacuum pressure source. The sample is placed in one cup, the filter assembly threaded onto the top of the cup, then the cup and filter are flipped, the receiving cup attached, vaccum tubing attached, vaccum pressure applied, and the sample is then pulled from the sample cup, through the filter assembly, and into the empty cup.
Systems include digestion cups and filtration assemblies. Tubing and vacuum source are not included.
Specifications
  • Pore Size (µm)0.7
  • Body MaterialPolypropylene, white caps
  • Funnel Size (mL)50
  • DescriptionFiltration Systems, Polypropylene, 0.7um, Glass Fiber Filter with Pre-Filter; 200/pk