Hand-Operated Vacuum/Pressure Pumps
Produce up to 25" Hg vacuum or test for pressure leaks
  • Pumps are sealed and self-lubricated for a long, maintenance-free life
  • Vacuum gauge is graduated in both centimeters and inches of mercury
  • Built-in trigger enables you to release vacuum without disconnecting the pump
  • Zinc alloy is perfect for rugged applications; PVC is lightweight and easy to handle
4,500.00 - ₹53,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hand-operated vacuum/pressure pump, 15 mL/stroke, zinc alloy body SKU 7930101 WW-79301-01 Mfr # MV8510
COMPARE
Hand-operated vacuum/pressure pump, 15 mL/stroke, zinc alloy body
₹19,100.00INR / Each
Hand-operated vacuum pump, 16 mL/stroke, PVC body WW-79301-11 Mfr # MV8010
COMPARE
Hand-operated vacuum pump, 16 mL/stroke, PVC body
₹17,500.00INR / Each
Hand-operated vacuum pump (without gauge), 32 mL/stroke, PVC body WW-79301-21 Mfr # MV8121
COMPARE
Hand-operated vacuum pump (without gauge), 32 mL/stroke, PVC body
₹33,000.00INR / Each
Repair kit for vacuum/pressure pump 79301-01 WW-79301-62 Mfr # MVM8900
COMPARE
Repair kit for vacuum/pressure pump 79301-01
₹6,500.00INR / Each
Repair kit for vacuum/pressure pump 79301-11 WW-79301-63 Mfr # MVM8901
COMPARE
Repair kit for vacuum/pressure pump 79301-11
₹4,500.00INR / Each
Repair kit for vacuum/pressure pump 79301-21 WW-79301-64 Mfr # MVM8902
COMPARE
Repair kit for vacuum/pressure pump 79301-21
₹53,000.00INR / Each