Environmental Express Autoclavable Mini pH Probe, Sealed/DJ/Glass; 180 mm
Representative image only.
Environmental Express Autoclavable Mini pH Probe, Sealed/DJ/Glass; 180 mm
Environmental Express | Item: 2904450
Get precise readings affordably
₹51,300.00INR / Each
₹51,300.00INR / Each
Buy It Together
Current Item
Environmental Express Autoclavable Mini pH Probe, Sealed/DJ/Glass; 180 mm
Environmental Express Autoclavable Mini pH Probe, Sealed/DJ/Glass; 180 mm
2904450
₹51,300.00INR / Each
+ =
Total Price 62700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Sealed
  • Autoclavable
  • 1 year warranty
More About this Item
Ideal for narrow crowded culture flasks and fermentors (6 mm dia). Double-junction reference offers stability and extended service life.
Length listed in table refers to probe immersion length, which is equal to housing length plus 70 mm.
1 Year Warranty
Specifications
  • ApplicationsAutoclavable, sealed
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Max Temperature (° C)130
  • DescriptionAutoclavable Mini pH Probe, Sealed/DJ/Glass; 180 mm