Cole-Parmer Reference filling solution for outer cell. High-temperature 3M KCl gel; 125 ml.
Representative image only.
Cole-Parmer Reference filling solution for outer cell. High-temperature 3M KCl gel; 125 ml.
Cole-Parmer | Item: 0566170
Get the most out of water analysis equipment
₹4,400.00INR / Each
₹4,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Reference filling solution for outer cell. High-temperature 3M KCl gel; 125 ml.
Cole-Parmer Reference filling solution for outer cell. High-temperature 3M KCl gel; 125 ml.
0566170
₹4,400.00INR / Each
+
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Transmitter
0580214
₹46,300.00INR / Each
+ + + =
Total Price 227300.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Replace or upgrade electrodes, probes, cells, and other accessories
  • 1 year warranty
More About this Item
Water analysis accessories for Cole-Parmer® meters. Many of these items are compatible with other brands of meter.
1 Year Warranty
Specifications
  • Compatible WithCole-Parmer autoclavable fermentation pH electrodes
  • DescriptionReference filling solution for outer cell. High-temperature 3M KCl gel; 125 ml.