EMD Millipore 1.17985.0001 Potassium Test Strips, 100
Representative image only.
EMD Millipore 1.17985.0001 Potassium Test Strips, 100
EMD Millipore | Mfr: 1.17985.0001| Item: 9957219
Use for quick spot checks to ensure water quality
Call For Price
Call For Price
Buy It Together
Current Item
EMD Millipore 1.17985.0001 Potassium Test Strips, 100
EMD Millipore 1.17985.0001 Potassium Test Strips, 100
9957219
Call For Price
+
Environmental Express Oakton pH Indicator Strips, 0 to 14, 100/pk
3585002
₹5,300.00INR / Pkg of 100
+ =
Total Price 14500.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.
Specifications
  • TestPotassium
  • Measurement Points0 to 1500 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionPotassium Test Strips, 100