Silver Lake Research WS-425D Drinking Water Test Kit , Single Use
Representative image only.
Silver Lake Research WS-425D Drinking Water Test Kit , Single Use
Silver Lake Research | Mfr: WS-425D| Item: 9953002
Lab-quality results in a simple-to-use, affordable kit
₹9,200.00INR / Each
₹9,200.00INR / Each
Buy It Together
Current Item
Silver Lake Research WS-425D Drinking Water Test Kit , Single Use
Silver Lake Research WS-425D Drinking Water Test Kit , Single Use
9953002
₹9,200.00INR / Each
+ =
Total Price 53400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Comprehensive test kits detect most common water contaminants
  • Compare your results against EPA-recommended levels
More About this Item
Fast, easy-to-use water test kits detect hazardous contaminants in municipal and private well drinking
water. Drinking water test kits indicate the presence of bacteria, pesticides, and lead. Kits also include readings for nitrate (0 to 50 ppm), nitrite (0 to 10 ppm), total chlorine (0 to 10 ppm), pH (6 to 10), and water hardness (0 to 425 ppm). Well water test kits add tests for contaminants commonly found in private well water: iron and copper (0 to 5 ppm). Order bacteria, pesticides, and lead stand-alone kits if less comprehensive tests are required. All kits are for one-time use.
Specifications
  • TestDrinking Water
  • Reagent PlatformStrips
  • Tests Per Kit1
  • DescriptionDrinking Water Test Kit , Single Use