EMD Millipore 1.10013.0001 Sulfite Test Strips, 10 to 400 ppm, 100/Pk
Representative image only.
EMD Millipore 1.10013.0001 Sulfite Test Strips, 10 to 400 ppm, 100/Pk
EMD Millipore | Mfr: 1.10013.0001| Item: 9957216
Use for quick spot checks to ensure water quality
₹39,600.00INR / Pkg of 100
₹39,600.00INR / Pkg of 100
Buy It Together
Current Item
EMD Millipore 1.10013.0001 Sulfite Test Strips, 10 to 400 ppm, 100/Pk
EMD Millipore 1.10013.0001 Sulfite Test Strips, 10 to 400 ppm, 100/Pk
9957216
₹39,600.00INR / Pkg of 100
+
Environmental Express Oakton pH Indicator Strips, 0 to 14, 100/pk
3585002
₹5,300.00INR / Pkg of 100
+ =
Total Price 54100.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.
Specifications
  • TestSulfite
  • Measurement Points0 to 400 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionSulfite Test Strips, 10 to 400 ppm, 100/Pk