EMD Millipore 1.10007.0001 Nitrite Test Strips, 2 to 80 ppm, 100/Pk
Representative image only.
EMD Millipore 1.10007.0001 Nitrite Test Strips, 2 to 80 ppm, 100/Pk
EMD Millipore | Mfr: 1.10007.0001| Item: 9957215
Use for quick spot checks to ensure water quality
₹43,200.00INR / Pkg of 100
₹43,200.00INR / Pkg of 100
Buy It Together
Current Item
EMD Millipore 1.10007.0001 Nitrite Test Strips, 2 to 80 ppm, 100/Pk
EMD Millipore 1.10007.0001 Nitrite Test Strips, 2 to 80 ppm, 100/Pk
9957215
₹43,200.00INR / Pkg of 100
+
Environmental Express Oakton pH Indicator Strips, 0 to 14, 100/pk
3585002
₹5,300.00INR / Pkg of 100
+ =
Total Price 57700.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.
Specifications
  • TestNitrite
  • Measurement Points0 to 80 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionNitrite Test Strips, 2 to 80 ppm, 100/Pk