Eldex Optos Model 3LI Piston Metering Pump, PEEK, 3/32" Piston Dia, 0.01 to 10 mL/min

Representative image only.

Eldex Optos Model 3LI Piston Metering Pump, PEEK, 3/32" Piston Dia, 0.01 to 10 mL/min

Eldex | Mfr: 5981| Item: 4853188

Compact pumps provide precise flow rate control and smooth fluid delivery at high pressures

₹515,000.00INR / Each

₹515,000.00INR / Each

Buy It Together

Current Item

Eldex Optos Model 3LI Piston Metering Pump, PEEK, 3/32" Piston Dia, 0.01 to 10 mL/min
Eldex Optos Model 3LI Piston Metering Pump, PEEK, 3/32" Piston Dia, 0.01 to 10 mL/min
4853188

₹515,000.00INR / Each
+ =
Total Price 693000.00 ADD 2 ITEMS TO CART
Product Description

Includes

Inlet and outlet fittings, 60 cm (23.5") L tubing, and 110/115 VAC power cord.

Key Features

  • Reciprocating piston design for precise flow control
  • High-resolution stepper motor minimizes pulsation
  • Control flow remotely via analog or RS-232 connection
  • Integrated piston wash extends piston seal life
  • Solvent-resistant membrane keypad
  • Use with liquids with temperatures ranging from 5 to 260°C (41 to 500°F)
  • Compact meter only measures 10 cm W x 24 cm D (4” x 9.5”), saving valuable benchtop space
  • PEEK liquid end cylinder
  • 1/2" stroke length
  • 3/32" piston diameter
  • Flow rate range from 0.01 to 10 mL/min
  • Manufacturer defect warranty

More About this Item

The Optos series metering pumps deliver high precision and reliability at high pressures. Achieve flow rates from 0.01 to 80 mL/min with ±0.3% reproducibility with Model 3 meters. The reciprocating piston offers precise flow control while an advanced software algorithms control high-resolution stepper motor to maximize flow output and minimize pulsation. Easily set flow rates using the up and down buttons on the solvent-resistant keypad. Remotely control your meter via an analog signal (4 to 20 mA or 0 to 5 VDC) or RS-232. Contact closures allow remote start and stop, flow output (0 to 5 VDC), and pump error output.

Select a pump that uses the upper half of the flow range listed for each pump. Piston cylinder comes available in two materials—316 stainless steel (SS) and PEEK. 316 SS offers excellent chemical and corrosion resistance; use PEEK for extremely corrosive fluids. A fully integrated piston wash system cleans out buffers or solutions which can crystallize and lead to abrasion of the piston seal, shortening the piston seal life.

A pressure differential of at least 25 psi is required on the outlet side of the pump for the valves to operate effectively.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Conformite Europeenne
Specifications
  • SeriesModel 3
  • Max Flow Rate (mL/min)10
  • Min Flow Rate (mL/min)0.01
  • Max Pressure (PSI)3000
  • Max Pressure (bar)206.8
  • Power (VAC)100 to 230
  • Power (Hz)50/60
  • InputRS-232, 0 to 5 VDC, 4 to 20 mA, remote start and stop
  • OutputFlow output (0 to 5 VDC), pump error output
  • Wetted MaterialsPEEK, sapphire, ruby, UHMW polyethylene, PCTFE
  • Inlet Connection1/4″-28 low pressure plastic fitting for use with 1/8″ plastic tubing
  • Outlet Connection1/4″-28 plastic fittings for use with 1/16″ tubing
  • ReproducibilityTypically ±0.3%
  • Min Temperature (° F)41
  • Max Temperature (° F)95
  • Min Temperature (° C)5
  • Max Temperature (° C)35
  • Max Viscosity (cP)500
  • Duty Cycle ContinuousYes
  • Tube Length, Inlet (in)23.6
  • Tube Length, Inlet (cm)60
  • Tube Od, Inlet (in)0.125
  • Width (in)4
  • Length (in)11
  • Height (in)9.25
  • Width (cm)10
  • Length (cm)28
  • Height (cm)23.5
  • DescriptionModel 3LI Piston Metering Pump, PEEK, 3/32" Piston Dia, 0.01 to 10 mL/min
  • WarrantyManufacturer defect
Reviews
Q&A