Eldex Optos Series Maintenance Kit for Pumps with PEEK Cylinder and 3/32" Dia Piston
Representative image only.
Eldex Optos Series Maintenance Kit for Pumps with PEEK Cylinder and 3/32" Dia Piston
Eldex | Mfr: 5923| Item: 4853194
Extend the life of your pump by keeping it well maintained
₹178,000.00INR / Each
₹178,000.00INR / Each
Product Description
Includes
Two UHMW PE piston seals, two check valve cartridges, retainer, filter, and tools.
Key Features
  • For pumps with 316 SS cylinder and 3/32" diameter piston
  • Manufacturer defect warranty
More About this Item
Optos series piston pumps are designed to provide years of trouble-free service. Ensure the longevity of your pump by performing a few simple maintenance procedures. Each maintenance kit contains piston seals, check valve cartridges, retainer, filter, and tools.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Conformite Europeenne
Specifications
  • Compatible WithPumps 48531-66, -69, -77, -80, and -88
  • DescriptionPiston Metering Pump Maintenance Kit for PEEK Cylinder and 3/32" Dia Piston
  • WarrantyManufacturer defect