Edwards Clean and Overhaul Kit for E1M18 and E2M18 Oil-Sealed Rotary Vane Vacuum Pumps
Representative image only.
Edwards Clean and Overhaul Kit for E1M18 and E2M18 Oil-Sealed Rotary Vane Vacuum Pumps
Edwards | Mfr: A36301131| Item: 7930063
Repair kits help prolong the life of your vacuum pump
₹105,000.00INR / Each
₹105,000.00INR / Each
Buy It Together
Current Item
Edwards Clean and Overhaul Kit for E1M18 and E2M18 Oil-Sealed Rotary Vane Vacuum Pumps
Edwards Clean and Overhaul Kit for E1M18 and E2M18 Oil-Sealed Rotary Vane Vacuum Pumps
7930063
₹105,000.00INR / Each
+ =
Total Price 1526000.00 ADD 2 ITEMS TO CART
Product Description
Includes
2x O-ring FKM GP 0.299 ID x .103 sec, 1x filter assy E2M 18/28, 2x O-ring NIT 1.109 ID x .139 sec, 2x O-ring NIT 99.5 ID x 3.0 sec, 1x dowty bonded seal MK3 2 BA, 8x O-ring NIT 6.6 ID x 2.4 sec, 1x O-ring NIT 15.6 ID x 2.4 sec, 1x O-ring NIT 13.6 ID x 2.4 sec, 2x dowty bonded seal MK A 1/8"BSP, 1x O-ring NIT 11.6 ID x 2.4 sec, 2x spirosel E1M18 FKM GP, 2x blade spring, 1x distributor rubber FKM, 3x blade springs, 1x coupling element EM18/28/30, 1x seal FKM GP 30.40.7 PLN, 1x O-ring FKM GP 194.5 ID x 3.0 sec, 3x valve pad, 1x oil drain plug RV, 1x O-ring NIT 27.5 ID x 3.0 sec, 1x felt pad, 1x label - Exhaust Warning, 1x gas ballast valve Peek450 CA30, 1x spring, 2x ballast gauze E1M18, 2x filter pad felt 1/4" thick, 2x oil filter seal, 2x oil drainplug 1/8" BSP, 1x oil box gasket E1M18, 2x spring, 1x O-ring FKM Gp 15.6 ID x 2.4 sec, 1x O-ring NIT 35.5 ID x 3.0 sec, 2x O-ring NIT 44.5 ID x 3.0 sec, 1x O-ring FKM GP 69.5 ID x 3.0 sec, 2x exhaust valve spring, 1x waved washer.
Key Features
  • Keep service parts handy to minimize downtime
  • Service kit for 79300-32, -33, -37, and -84
  • 1 year warranty
More About this Item
Genuine parts provide a perfect fit ensuring reliable performance.
The clean and overhaul kit contains all of the springs, gaskets, seals, and other components required for routine maintenance and whenever the pump is dismantled.
Fluoroelastomer clean and overhaul kit works with the following models:
E1M18 rotary vane pumps
E2M18 rotary vane pumps
1 Year Warranty
Specifications
  • Compatible WithE1M18 and E2M18 oil-sealed rotary vane vacuum pumps
  • TypeClean and Overhaul Kit
  • DescriptionClean and Overhaul Kit for E1M18 and E2M18 Oil-Sealed Rotary Vane Vacuum Pumps