Edwards C105-14-645 Vacuum Pump Fitting, Aluminum Adapter, NW 25 x 3/8" Hose Barb
Representative image only.
Edwards C105-14-645 Vacuum Pump Fitting, Aluminum Adapter, NW 25 x 3/8" Hose Barb
Edwards | Mfr: C105-14-645| Item: 3140169
Use with a clamp and co-seal or centering ring
₹10,700.00INR / Each
₹10,700.00INR / Each
Buy It Together
Current Item
Edwards C105-14-645 Vacuum Pump Fitting, Aluminum Adapter, NW 25 x 3/8" Hose Barb
Edwards C105-14-645 Vacuum Pump Fitting, Aluminum Adapter, NW 25 x 3/8" Hose Barb
3140169
₹10,700.00INR / Each
+ =
Total Price 16800.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Aluminum construction
  • NW 25 flange x 3/8" hose barb
More About this Item
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring.
Specifications
  • Fitting MaterialAluminum
  • Connection TypeVacuum Flange x Barb
  • Barb Size / Tubing Id (Port 2) (in)3/8
  • Flange Size (Port 1)NW 25
  • Min Temperature (° C)-10
  • Max Temperature (° C)150
  • Max Vacuum (in Hg)44.29
  • DescriptionVacuum Pump Fittings, aluminum adapter, NW 25 by 1/2 to 3/8" hose barb