Edwards C105-02-203 Vacuum Pump Fittings, 316L SS adapter, NW 25 to 1" OD tubing
Representative image only.
Edwards C105-02-203 Vacuum Pump Fittings, 316L SS adapter, NW 25 to 1" OD tubing
Edwards | Mfr: C105-02-203| Item: 3140145
Use with a clamp and co-seal or centering ring
₹56,300.00INR / Each
₹56,300.00INR / Each
Buy It Together
Current Item
Edwards C105-02-203 Vacuum Pump Fittings, 316L SS adapter, NW 25 to 1" OD tubing
Edwards C105-02-203 Vacuum Pump Fittings, 316L SS adapter, NW 25 to 1" OD tubing
3140145
₹56,300.00INR / Each
+ =
Total Price 62400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring.
Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeVacuum Flange x Compression
  • Compression Size / Tube Od (Port 2) (in)1
  • Flange Size (Port 1)NW 25
  • DescriptionVacuum Pump Fittings, 316L SS adapter, NW 25 to 1" OD tubing