Edwards C105-01-414 Vacuum Pump Fittings, 316L SS adapter, NW 25 to 3/4"BSP(M)

Representative image only.

Edwards C105-01-414 Vacuum Pump Fittings, 316L SS adapter, NW 25 to 3/4"BSP(M)

Edwards | Mfr: C105-01-414| Item: 7930250

Use with a clamp and co-seal or centering ring

₹35,400.00INR / Each

₹35,400.00INR / Each

Buy It Together

Current Item

Edwards C105-01-414 Vacuum Pump Fittings, 316L SS adapter, NW 25 to 3/4"BSP(M)
Edwards C105-01-414 Vacuum Pump Fittings, 316L SS adapter, NW 25 to 3/4"BSP(M)
7930250

₹35,400.00INR / Each
+ =
Total Price 41500.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring.

1 Year Warranty
Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeVacuum Flange x BSP(M)
  • Thread size BSP(M) (port 2) (in)3/4
  • Flange Size (Port 1)NW 25
  • DescriptionVacuum Pump Fittings, 316L SS adapter, NW 25 to 3/4"BSP(M)
  • Warranty1 year
Reviews
Q&A