Edwards C105-01-222 Vacuum Pump Fittings, aluminum adapter, NW 25 to 1/4"NPT(F)

Representative image only.

Edwards C105-01-222 Vacuum Pump Fittings, aluminum adapter, NW 25 to 1/4"NPT(F)

Edwards | Mfr: C105-01-222| Item: 3140135

Use with a clamp and co-seal or centering ring

₹18,100.00INR / Each

₹18,100.00INR / Each

Buy It Together

Current Item

Edwards C105-01-222 Vacuum Pump Fittings, aluminum adapter, NW 25 to 1/4"NPT(F)
Edwards C105-01-222 Vacuum Pump Fittings, aluminum adapter, NW 25 to 1/4"NPT(F)
3140135

₹18,100.00INR / Each
+ =
Total Price 24200.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring.

Specifications
  • Fitting MaterialAluminum
  • Connection TypeVacuum Flange x NPT(F)
  • Thread size NPT(F) (port 2) (in)1/4
  • Flange Size (Port 1)NW 25
  • DescriptionVacuum Pump Fittings, aluminum adapter, NW 25 to 1/4"NPT(F)
Reviews
Q&A