Edwards C105-01-109 Vacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"VCR(F)

Representative image only.

Edwards C105-01-109 Vacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"VCR(F)

Edwards | Mfr: C105-01-109| Item: 3140121

Use with a clamp and co-seal or centering ring

₹46,900.00INR / Each

₹46,900.00INR / Each

Buy It Together

Current Item

Edwards C105-01-109 Vacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"VCR(F)
Edwards C105-01-109 Vacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"VCR(F)
3140121

₹46,900.00INR / Each
+ =
Total Price 53000.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring.

Specifications
  • Fitting MaterialStainless Steel
  • Connection TypeVacuum Flange x VCR
  • Flange Size (Port 1)NW 16
  • Flange Size (Port 2)0.25" VCR
  • DescriptionVacuum Pump Fittings, 316L SS adapter, NW 16 to 1/4"VCR(F)
Reviews
Q&A