Edwards C105-01-105 Vacuum Pump Fitting, Aluminum Adapter, NW 16 x 1/4" NPT(F)

Representative image only.

Edwards C105-01-105 Vacuum Pump Fitting, Aluminum Adapter, NW 16 x 1/4" NPT(F)

Edwards | Mfr: C105-01-105| Item: 3140109

Use with a clamp and co-seal or centering ring

₹15,000.00INR / Each

₹15,000.00INR / Each

Buy It Together

Current Item

Edwards C105-01-105 Vacuum Pump Fitting, Aluminum Adapter, NW 16 x 1/4" NPT(F)
Edwards C105-01-105 Vacuum Pump Fitting, Aluminum Adapter, NW 16 x 1/4" NPT(F)
3140109

₹15,000.00INR / Each
+ =
Total Price 21100.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Aluminum construction
  • NW 16 to 1/4" NPT(F)

More About this Item

Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring.

Specifications
  • Fitting MaterialAluminum
  • Connection TypeVacuum Flange x NPT(F)
  • Thread size NPT(F) (port 2) (in)1/4
  • Flange Size (Port 1)NW 16
  • DescriptionVacuum Pump Fittings, aluminum adapter, NW 16 to 1/4"NPT(F)
Reviews
Q&A