Dynalon Air or Water-Driven Magnetic Stirrer, 0-800 rpm, 500 mL Capacity

Representative image only.

Dynalon Air or Water-Driven Magnetic Stirrer, 0-800 rpm, 500 mL Capacity

Dynalon | Mfr: 204015| Item: 0463400

Corrosion-proof plastic eliminates shock hazard in dangerous environments.

₹41,800.00INR / Each

for an additional
₹41,400.00INR / Each
for an additional
₹29,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹41,800.00INR / Each

Buy It Together

Current Item

Dynalon Air or Water-Driven Magnetic Stirrer, 0-800 rpm, 500 mL Capacity
Dynalon Air or Water-Driven Magnetic Stirrer, 0-800 rpm, 500 mL Capacity
0463400

₹41,800.00INR / Each
+
InnoCal NIST-Traceable Calibration; Stirrer Only
1705004

₹29,300.00INR / Each
=
Total Price 71100.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 1 year warranty

More About this Item

Use stirrer in hazardous locations or on your benchtop. Position stirrer off your benchtop by connecting it to a 1/2'''' or 3/8'''' dia support rod with the built-in ring clamp. Inlet ID measures 4.1 mm; outlet ID measures 9.1 mm.
Attach stirrer to any air pressure line or water source—stirs 500 mL of water at 800 rpm with air at 8 psi or with water at 12 psi. What''s included: 6-ft of vinyl inlet and 1-ft outlet tubing for easy hookup to your air or water source.

1 Year Warranty
Specifications
  • Top Plate Diameter (in)5
  • Top Plate Diameter (cm)12.7
  • Top Plate MaterialPolypropylene
  • Low Speed (rpm)0
  • High Speed (rpm)800
  • No. Of Stirring Positions1
  • Max Stirring Volume (Liters)0.5
  • Length (in)5
  • Height (in)2 5/8
  • Length (cm)12.7
  • Height (cm)6.7
  • Diameter (in)5
  • Diameter (cm)12.7
  • DescriptionAir or Water-Driven Magnetic Stirrer, 0-800 rpm, 500 mL Capacity
  • Warranty1 year
Reviews
Q&A