Drager Diffusion Tube, Ammonia 20/a-D, 20 to 1500 ppm at 1 hour

Representative image only.

Drager Diffusion Tube, Ammonia 20/a-D, 20 to 1500 ppm at 1 hour

Drager | Mfr: 8101301| Item: 8651552

Direct read tubes identify workers exposure to chemicals

₹44,700.00INR / Box of 10

₹44,700.00INR / Box of 10

Buy It Together

Current Item

Drager Diffusion Tube, Ammonia 20/a-D, 20 to 1500 ppm at 1 hour
Drager Diffusion Tube, Ammonia 20/a-D, 20 to 1500 ppm at 1 hour
8651552

₹44,700.00INR / Box of 10
+
Drager Diffusion Tube Holder with clip, pack of 3
8651550

₹27,500.00INR / Pkg of 3
=
Total Price 72200.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Read exposure levels easily but he markings on the side of the tube
  • 2 year warranty

More About this Item

Direct-reading diffusion tubes are used specifically for personal exposure monitoring. These tubes do not require the use of a gas detector pump because the contaminant molecules pass into the tube by diffusion processes and in this manner are brought into contact with the reagent systems. Exposure levels are easily read by the markings on the side of the tube.

Two year warranty
Specifications
  • Gas MeasuredAmmonia 20/a-D
  • Range1 hr: 20 to 1500 ppm, 4 hr: 2.5 to 200 ppm
  • DescriptionDiffusion Tube, Ammonia 20/a-D, 20 to 1500 ppm at 1 hour
  • Warranty2 year
Reviews
Q&A