Drager Diffusion Tube Holder with clip, pack of 3
Representative image only.
Drager Diffusion Tube Holder with clip, pack of 3
Drager | Mfr: 6733014| Item: 8651550
Direct read tubes identify workers exposure to chemicals
₹27,500.00INR / Pkg of 3
₹27,500.00INR / Pkg of 3
Product Description
Key Features
  • Read exposure levels easily but he markings on the side of the tube
More About this Item
Direct-reading diffusion tubes are used specifically for personal exposure monitoring. These tubes do not require the use of a gas detector pump because the contaminant molecules pass into the tube by diffusion processes and in this manner are brought into contact with the reagent systems. Exposure levels are easily read by the markings on the side of the tube.
Specifications
  • DescriptionDiffusion Tube Holder with clip, pack of 3